Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mall | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145532.3 | NP_663507.1 | 2 | 235 | ||
GenBank mRNA | AK078971.1 | - | 2 | 236 | ||
AK163867.1 | - | 2 | 236 | |||
BC012256.1 | - | 2 | 270 | 4205488 |