Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177545.5 | NP_808213.2 | 5 | 1185 | ||
GenBank mRNA | AF481860.1 | - | 5 | 1101 | ||
AK135230.1 | - | 5 | 1146 | |||
AK167743.1 | - | 6 | 999 | |||
BC138519.1 | - | 4 | 927 | 8861539 | ||
BC138520.1 | - | 4 | 927 | 8861540 | ||
BU610139.1 | - | 1 | 31 |