Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172685.3 | NP_766273.1 | 6 | 852 | ||
GenBank mRNA | AK032806.1 | - | 6 | 853 | ||
AK146034.1 | - | 6 | 851 | |||
AK167778.1 | - | 6 | 843 | |||
AK168090.1 | - | 6 | 843 | |||
AK170598.1 | - | 6 | 844 | |||
BC022637.1 | - | 1 | 67 | |||
BC055369.1 | - | 6 | 841 | 30055307 |