Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163283.1 | NP_001156755.1 | 2 | 1952 | ||
NM_001163284.1 | NP_001156756.1 | 2 | 2847 | |||
NM_173399.3 | NP_775575.2 | 2 | 1575 | |||
GenBank mRNA | AK031172.1 | - | 2 | 1507 | ||
AK129124.1 | - | 2 | 2410 | |||
AK160003.1 | - | 2 | 2564 | |||
AY635051.1 | - | 2 | 2847 | |||
BC054551.1 | - | 2 | 1433 | 6400036 | ||
BC057545.1 | - | 2 | 2847 | 6405028 |