Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam206a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081420.1 | NP_001074889.1 | 5 | 678 | ||
GenBank mRNA | AK052647.1 | - | 5 | 493 | ||
AK164596.1 | - | 5 | 491 | |||
BC026590.1 | - | 5 | 657 | 4973099 | ||
BC048548.1 | - | 5 | 629 | 6590165 | ||
BC062190.1 | - | 6 | 659 |