Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172694.2 | NP_766282.1 | 5 | 1316 | ||
GenBank mRNA | AK035987.1 | - | 5 | 1317 | ||
AK039319.1 | - | 5 | 1317 | |||
AK122378.1 | - | 1307 | ||||
BC088994.1 | - | 5 | 1225 | 6734479 | ||
BC138128.1 | - | 5 | 1255 | 8861147 | ||
BC138129.1 | - | 5 | 1255 | 8861148 |