Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146159.1 | NP_666271.1 | 2 | 441 | ||
GenBank mRNA | AK049414.1 | - | 1 | 1411 | ||
BC023882.1 | - | 2 | 473 | 5354698 | ||
BC025497.1 | - | 2 | 459 | 5320270 | ||
BC027393.1 | - | 2 | 441 | 4948448 |