Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139064.2 | NP_620703.1 | 6 | 1171 | ||
GenBank mRNA | AJ304865.1 | - | 6 | 1171 | ||
AK160165.1 | - | 6 | 1129 | |||
BC052083.1 | - | 7 | 1217 | 6404610 |