Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172711.2 | NP_766299.1 | 6 | 679 | ||
GenBank mRNA | AK032597.1 | - | 5 | 1687 | ||
AK047411.1 | - | 6 | 548 | |||
AK053420.1 | - | 6 | 653 | |||
AK084627.1 | - | 6 | 660 | |||
AK140534.1 | - | 6 | 638 | |||
AK160956.1 | - | 6 | 698 | |||
AK165012.1 | - | 6 | 668 | |||
BC057962.1 | - | 6 | 633 | 5353891 |