Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172719.2 | NP_766307.2 | 50 | 6785 | ||
GenBank mRNA | AB093217.1 | - | 26 | 3978 | ||
AK077013.1 | - | 14 | 2104 | |||
AK090137.1 | - | 50 | 6782 | |||
AK147257.1 | - | 50 | 6778 | |||
BC038407.1 | - | 3 | 522 | |||
BC056933.1 | - | 16 | 2417 | |||
BC068244.1 | - | 6 | 1039 | |||
BC150735.1 | - | 50 | 6788 | 9087646 |