Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 6 | 892 | ||
GenBank mRNA | AK040924.1 | - | 4 | 444 | ||
AK146431.1 | - | 6 | 770 | |||
AK152402.1 | - | 6 | 770 | |||
AK153787.1 | - | 6 | 765 | |||
AK181051.1 | - | 1 | 108 | |||
AK218835.1 | - | 3 | 238 | |||
BC050232.1 | - | 4 | 489 | |||
BC057548.1 | - | 6 | 747 | |||
BC082786.1 | - | 6 | 865 | |||
BC099419.1 | - | 6 | 824 | |||
BC144755.1 | - | 4 | 423 |