Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Radil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289588.1 | NP_001276517.1 | 2 | 654 | ||
NM_178702.4 | NP_848817.2 | 2 | 654 | |||
GenBank mRNA | AK038570.1 | - | 2 | 587 | ||
AK052964.1 | - | 2 | 565 | |||
AK083410.1 | - | 2 | 672 | |||
AK162917.1 | - | 2 | 634 | |||
AK163852.1 | - | 2 | 590 | |||
AK173277.1 | - | 2 | 618 | |||
BC056483.1 | - | 2 | 606 | 5696312 |