Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172146.1 | NP_001165617.1 | 2 | 413 | ||
NM_146165.2 | NP_666277.1 | 2 | 293 | |||
GenBank mRNA | BC024410.1 | - | 2 | 386 | 4954972 | |
BC024480.1 | - | 2 | 284 | 4977193 | ||
BC026958.1 | - | 2 | 352 | 4013019 | ||
BC026972.1 | - | 2 | 366 | 5371230 | ||
CF732613.1 | - | 2 | 335 | |||
CJ051555.1 | - | 2 | 285 |