Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290377.1 | NP_001277306.1 | 4 | 652 | ||
NM_175632.3 | NP_783440.1 | 4 | 670 | |||
GenBank mRNA | AF391159.1 | - | 4 | 628 | ||
AF391160.1 | - | 4 | 652 | |||
AF391161.1 | - | 4 | 646 | |||
BC137776.1 | - | 4 | 597 | 8860795 |