Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172739.4 | NP_766327.3 | 2 | 4051 | ||
GenBank mRNA | AK136766.1 | - | 1 | 725 | ||
AK147326.1 | - | 2 | 4051 | |||
AK147688.1 | - | 2 | 4051 | |||
AK161796.1 | - | 2 | 3646 | |||
AK173242.1 | - | 1 | 2853 | |||
BC035541.1 | - | 1 | 114 | |||
BC139459.1 | - | 3 | 3829 | 8862481 | ||
BC139461.1 | - | 3 | 3688 | 8862483 |