Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172739.4 | NP_766327.3 | 4, 5 | 4628 | ||
GenBank mRNA | AK136766.1 | - | 3, 4 | 1302 | ||
AK147326.1 | - | 4, 5 | 4628 | |||
AK147688.1 | - | 4, 5 | 4628 | |||
AK173242.1 | - | 3, 4 | 3430 | |||
BC035541.1 | - | 3, 4 | 691 | |||
BC139459.1 | - | 5, 6 | 4406 | 8862481 | ||
BC139461.1 | - | 5, 6 | 4265 | 8862483 |