Gene Symbol: Akap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029332.1 | NP_083608.1 | 15, 16 | 5261 | ||
GenBank mRNA | AK135576.1 | - | 7, 8 | 1069 | ||
AK149507.1 | - | 8, 9 | 2560 | |||
AY033771.1 | - | 10, 11 | 2587 |