Gene Symbol: Akap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029332.1 | NP_083608.1 | 11 | 4614 | ||
GenBank mRNA | AK135576.1 | - | 4 | 476 | ||
AK149507.1 | - | 5 | 1967 | |||
AK162999.1 | - | 5 | 1066 | |||
AK172678.1 | - | 3 | 402 | |||
AY033771.1 | - | 5 | 1874 |