Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201352.2 | NP_958740.2 | 7 | 1243 | ||
GenBank mRNA | AK141189.1 | - | 7 | 1276 | ||
AK154771.1 | - | 7 | 1380 | |||
BC024955.1 | - | 6 | 529 | 4953174 | ||
BC026428.1 | - | 3 | 176 | 4224574 | ||
BC082585.1 | - | 7 | 1243 | 30608279 |