Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153525.5 | NP_705745.3 | 7 | 976 | ||
GenBank mRNA | AK029327.1 | - | 7 | 977 | ||
AK033997.1 | - | 7 | 1006 | |||
AK035266.1 | - | 8 | 821 | |||
AK035365.1 | - | 5 | 554 | |||
AK038953.1 | - | 7 | 936 | |||
AK046140.1 | - | 2 | 223 | |||
AK053588.1 | - | 7 | 975 | |||
AK075726.1 | - | 7 | 974 | |||
AK078357.1 | - | 7 | 977 | |||
AK090100.1 | - | 7 | 978 | |||
AK139961.1 | - | 7 | 977 | |||
AK150353.1 | - | 7 | 1032 | |||
AK153048.1 | - | 7 | 1032 | |||
AK165226.1 | - | 7 | 977 | |||
AK166845.1 | - | 6 | 792 | |||
AK167021.1 | - | 7 | 878 | |||
AK172876.1 | - | 7 | 995 | |||
BC026515.1 | - | 7 | 1000 | 4910818 | ||
BC027103.1 | - | 7 | 977 | 5360942 | ||
BC086759.1 | - | 1429 |