Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145585.2 | NP_663560.1 | 3, 4 | 719 | ||
GenBank mRNA | AK032078.1 | - | 3, 4 | 695 | ||
AK152912.1 | - | 3, 4 | 692 | |||
AK178112.1 | - | 1, 2 | 145 | |||
BC004776.1 | - | 3, 4 | 686 | 3257051 |