Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139149.2 | NP_631888.1 | 4 | 318 | ||
GenBank mRNA | AF224264.1 | - | 4 | 243 | ||
AK032815.1 | - | 4 | 307 | |||
AK038226.1 | - | 4 | 304 | |||
AK040203.1 | - | 1 | 92 | |||
AK086587.1 | - | 1 | 83 | |||
AK132141.1 | - | 311 | ||||
AK134491.1 | - | 4 | 311 | |||
AK139960.1 | - | 4 | 317 | |||
AK145582.1 | - | 4 | 308 | |||
AK146198.1 | - | 4 | 311 | |||
AK149118.1 | - | 4 | 310 | |||
AK172554.1 | - | 4 | 303 | |||
BC011078.1 | - | 4 | 301 | |||
BC040827.1 | - | 4 | 273 | 4503820 | ||
BC058247.1 | - | 4 | 293 | 6314291 | ||
BY269119.1 | - | 4 | 341 |