Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173410.3 | NP_775586.1 | 3 | 1350 | ||
GenBank mRNA | AK036100.1 | - | 3 | 1353 | ||
AK042755.1 | - | 3 | 452 | |||
BC119380.1 | - | 3 | 1208 | 8734129 | ||
BC120647.1 | - | 3 | 1208 | 40129570 |
Set Membership: |
![]() |