Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172760.3 | NP_766348.3 | 19, 20 | 2097 | ||
GenBank mRNA | AK037067.1 | - | 19, 20 | 2090 | ||
BC018516.1 | - | 17, 18 | 1986 | 3155913 | ||
BC026617.1 | - | 14, 15 | 2042 | |||
BC058752.1 | - | 19, 20 | 2080 | 6485393 | ||
BC138887.1 | - | 19, 20 | 2054 | 8861909 |