Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175646.3 | NP_783577.2 | 4 | 581 | ||
GenBank mRNA | AK079533.1 | - | 4 | 680 | ||
AK085226.1 | - | 4 | 562 | |||
BC094379.1 | - | 4 | 597 | 30541506 | ||
BC132373.1 | - | 3 | 506 | 40130650 | ||
BC138333.1 | - | 3 | 506 | 8861353 |