Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172769.2 | NP_766357.1 | 6 | 915 | ||
GenBank mRNA | AB016248.1 | - | 6 | 901 | ||
AK043825.1 | - | 6 | 874 | |||
AK077670.1 | - | 6 | 873 | |||
AK154309.1 | - | 6 | 862 | |||
BC024132.1 | - | 6 | 864 | 5101986 |