Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145223.2 | NP_660258.2 | 10 | 5340 | ||
GenBank mRNA | AF425257.1 | - | 10 | 5341 |