Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153403.2 | NP_700452.2 | 3 | 241 | ||
GenBank mRNA | AB081471.1 | - | 3 | 241 | ||
BC129914.1 | - | 3 | 241 | 40111808 | ||
BC129915.1 | - | 3 | 237 |