Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079847.2 | NP_001073316.1 | 22 | 2081 | ||
NM_001079848.2 | NP_001073317.1 | 21 | 2051 | |||
NM_001079857.2 | NP_001073326.1 | 22 | 2090 | |||
NM_001290445.1 | NP_001277374.1 | 22 | 2084 | |||
NM_001290446.1 | NP_001277375.1 | 23 | 2123 | |||
NM_178712.4 | NP_848827.1 | 23 | 2132 | |||
GenBank mRNA | AF538952.1 | - | 23 | 2097 | ||
AF538955.1 | - | 22 | 2049 | |||
AF538956.1 | - | 23 | 2088 | |||
AF538957.1 | - | 21 | 2016 | |||
AK041291.1 | - | 24 | 2321 | |||
AK046871.1 | - | 22 | 2057 | |||
BC115874.1 | - | 21 | 2026 | 40086356 | ||
BC116644.1 | - | 22 | 2077 | 40086357 |
Set Membership: |
![]() |