Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033330.3 | NP_001028502.1 | 7 | 1012 | ||
NM_001290427.1 | NP_001277356.1 | 5 | 623 | |||
NM_001290428.1 | NP_001277357.1 | 4 | 556 | |||
GenBank mRNA | AK147370.1 | - | 7 | 1013 | ||
AK147440.1 | - | 4 | 557 | |||
BC139429.1 | - | 5 | 468 | 8862451 |