Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080793.2 | NP_001074262.1 | 9, 10 | 867 | ||
NM_001286207.1 | NP_001273136.1 | 9, 10 | 840 | |||
NM_011791.3 | NP_035921.2 | 9, 10 | 957 | |||
GenBank mRNA | AB020983.2 | - | 9, 10 | 939 | ||
AK087934.1 | - | 9, 10 | 932 | |||
AK132471.1 | - | 8, 9 | 863 | |||
AK145576.1 | - | 9, 10 | 832 | |||
AK145790.1 | - | 9, 10 | 844 | |||
AK145967.1 | - | 9, 10 | 811 | |||
AK146254.1 | - | 9, 10 | 869 | |||
AK146599.1 | - | 9, 10 | 841 | |||
AK146756.1 | - | 9, 10 | 832 | |||
AK146938.1 | - | 9, 10 | 934 | |||
AK183467.1 | - | 4, 5 | 297 | |||
BC012957.1 | - | 9, 10 | 944 | 4211433 |