Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080793.2 | NP_001074262.1 | 7, 8 | 706 | ||
NM_001286207.1 | NP_001273136.1 | 7, 8 | 679 | |||
NM_011791.3 | NP_035921.2 | 7, 8 | 796 | |||
GenBank mRNA | AB020983.2 | - | 7, 8 | 778 | ||
AK087934.1 | - | 7, 8 | 771 | |||
AK145576.1 | - | 7, 8 | 671 | |||
AK145790.1 | - | 7, 8 | 683 | |||
AK145967.1 | - | 7, 8 | 650 | |||
AK146254.1 | - | 7, 8 | 708 | |||
AK146599.1 | - | 7, 8 | 680 | |||
AK146756.1 | - | 7, 8 | 671 | |||
AK146938.1 | - | 7, 8 | 773 | |||
AK178767.1 | - | 3, 4 | 252 | |||
AK183467.1 | - | 2, 3 | 136 | |||
AK218371.1 | - | 3, 4 | 252 | |||
BC012957.1 | - | 7, 8 | 783 | 4211433 |