Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172805.3 | NP_766393.2 | 6 | 1421 | ||
GenBank mRNA | AK032438.1 | - | 6 | 1422 | ||
BC066138.1 | - | 6 | 763 | 30094199 |