Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 11 | 1429 | ||
GenBank mRNA | AB025269.1 | - | 10 | 996 | ||
AK154243.1 | - | 11 | 1428 | |||
AK172042.1 | - | 11 | 1442 | |||
AK208654.1 | - | 3 | 239 | |||
AK210574.1 | - | 4 | 309 | |||
AK216106.1 | - | 3 | 235 | |||
AK219454.1 | - | 3 | 226 | |||
BC057007.1 | - | 11 | 1324 | 6816943 | ||
BC062971.1 | - | 11 | 1429 | 6415619 |