Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175499.4 | NP_780708.1 | 2 | 1221 | ||
GenBank mRNA | AK029275.1 | - | 2 | 1144 | ||
AK044796.1 | - | 2 | 1222 | |||
BC138664.1 | - | 2 | 738 | 8861685 | ||
BC145927.1 | - | 2 | 738 | 40131230 |