Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032540.3 | NP_115929.3 | 9, 10 | 1269 | ||
GenBank mRNA | AF252870.1 | - | 9, 10 | 1241 | ||
AK028430.1 | - | 9, 10 | 1243 | |||
BC099961.1 | - | 9, 10 | 1267 | 30035694 |