Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181543.1 | NP_853521.1 | 1 | 637 | ||
GenBank mRNA | AJ564167.1 | - | 1 | 637 | ||
AK048591.1 | - | 1 | 295 | |||
AY351677.1 | - | 1 | 584 | |||
BC120520.1 | - | 1 | 617 | 40129443 | ||
BC120526.1 | - | 1 | 617 | 40129449 |
Set Membership: |
![]() |