Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153405.2 | NP_700454.1 | 6 | 1118 | ||
GenBank mRNA | AB036992.1 | - | 6 | 1034 | ||
AK080420.1 | - | 6 | 1092 | |||
AK089975.1 | - | 6 | 1011 | |||
AK090084.1 | - | 6 | 1087 | |||
AK140861.1 | - | 6 | 1119 | |||
AY152391.1 | - | 6 | 1118 | |||
BC057890.1 | - | 6 | 1029 | 4979982 |