Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp385b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113399.1 | NP_001106870.1 | 6 | 916 | ||
NM_001113400.1 | NP_001106871.1 | 6 | 938 | |||
NM_178723.6 | NP_848838.2 | 8 | 1448 | |||
GenBank mRNA | AK046798.1 | - | 8 | 1449 | ||
AK050293.1 | - | 6 | 939 | |||
AK078551.1 | - | 6 | 893 | |||
AK079322.1 | - | 8 | 1333 | |||
AK149352.1 | - | 8 | 1429 | |||
BC132352.1 | - | 6 | 1027 | 40130629 | ||
BC138239.1 | - | 6 | 1027 | 8861258 |