Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080944.3 | NP_001074413.2 | 13 | 1200 | ||
GenBank mRNA | AK040903.1 | - | 14 | 1369 | ||
AK086607.1 | - | 13 | 1234 | |||
AK155448.1 | - | 13 | 1210 | |||
AK169967.1 | - | 13 | 1223 | |||
AK173307.1 | - | 13 | 1200 |