Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177732.4 | NP_808400.1 | 1 | 129 | ||
GenBank mRNA | AK049180.1 | - | 1 | 130 | ||
AK049462.1 | - | 1 | 49 | |||
AK052968.1 | - | 1 | 49 | |||
AK140305.1 | - | 1 | 421 | |||
AK164517.1 | - | 1 | 49 | |||
AK169775.1 | - | 1 | 31 | |||
AK220330.1 | - | 1 | 39 |