Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177732.4 | NP_808400.1 | 3 | 306 | ||
GenBank mRNA | AK037934.1 | - | 1 | 541 | ||
AK049180.1 | - | 3 | 307 | |||
AK049462.1 | - | 3 | 226 | |||
AK052968.1 | - | 3 | 226 | |||
AK140305.1 | - | 3 | 598 | |||
AK164517.1 | - | 3 | 226 | |||
AK169775.1 | - | 3 | 208 | |||
AK220330.1 | - | 3 | 216 | |||
BC115984.1 | - | 4 | 207 | 40105299 |