Search Thermo Fisher Scientific
Gene Symbol: Tecrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153801.3 | NP_722496.2 | 7, 8 | 910 | ||
GenBank mRNA | AF548365.1 | - | 7, 8 | 911 | ||
AK052276.1 | - | 7, 8 | 749 | |||
AK052284.1 | - | 7, 8 | 913 | |||
AK052302.1 | - | 7, 8 | 615 | |||
AK052378.1 | - | 7, 8 | 673 | |||
BC107366.1 | - | 7, 8 | 823 | 40056219 | ||
BC107367.1 | - | 7, 8 | 823 | 40056223 |