Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081342.1 | NP_001074811.1 | 19 | 2406 | ||
GenBank mRNA | AK054021.1 | - | 4 | 253 | ||
AK153762.1 | - | 19 | 2409 | |||
AY255618.1 | - | 4 | 291 | |||
BC151161.1 | - | 19 | 2108 | 9088050 | ||
BC157994.1 | - | 18 | 2012 | 9007307 | ||
BC158001.1 | - | 18 | 2012 | 9007310 |
Set Membership: |
G-protein coupled receptor |