Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172298.2 | NP_758502.1 | 2 | 1209 | ||
GenBank mRNA | AK041793.1 | - | 1 | 95 | ||
AK220396.1 | - | 2 | 1080 | |||
AY063491.1 | - | 2 | 1209 | |||
BC150674.1 | - | 2 | 1152 | 9087585 |