Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177382.4 | NP_796356.2 | 2 | 269 | ||
GenBank mRNA | AY323818.1 | - | 2 | 269 | ||
BC108961.1 | - | 2 | 269 | 40046632 |