Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172913.3 | NP_766501.2 | 3, 4 | 712 | ||
GenBank mRNA | AK082602.1 | - | 3, 4 | 710 | ||
BC052044.1 | - | 3, 4 | 438 | 5708447 | ||
EF410108.1 | - | 576 |