Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291211.1 | NP_001278140.1 | 3 | 412 | ||
NM_153594.3 | NP_705822.1 | 3 | 412 | |||
GenBank mRNA | AB182640.1 | - | 3 | 353 | ||
AK031335.1 | - | 3 | 367 | |||
AK088800.1 | - | 3 | 351 | |||
AK146080.1 | - | 3 | 359 | |||
BC031360.1 | - | 2 | 229 | 5359242 | ||
BC040385.1 | - | 3 | 363 | 3499499 |