Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145374.2 | NP_663349.2 | 3 | 1131 | ||
GenBank mRNA | BC020002.1 | - | 3 | 1129 | 3584335 | |
BC025652.1 | - | 1 | 199 | |||
CX565910.1 | - | 1 | 319 |