Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146304.2 | NP_666416.2 | 1 | 416 | ||
GenBank mRNA | BC106795.1 | - | 1 | 416 | 40042190 | |
BC126866.1 | - | 1 | 410 | |||
BC126867.1 | - | 1 | 411 | 40042921 |